Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCAM1 Recombinant Protein fused to murine IgG2a Fc purifird from CHO cells

Catalog No. 89953677 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-677 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-677 Supplier Abnova™ Supplier No. P4935
Add to Cart
Add to Cart

Used for SDS-PAGE

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 4684
Molecular Weight (g/mol) 67.8kDa
Name NCAM1 (Human) Recombinanat Protein
Preparation Method Mammalian cell (CHO) expression system
Purification Method Affinity and size purification
Quantity 25 μg
Storage Requirements Store at 4°C. Open under aseptic conditions. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias CD56/MSK39/NCAM
Common Name NCAM1
Gene Symbol NCAM1
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.