Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCBP1 Full-length ORF (NP_002477.1, 1 a.a. - 790 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-690 25 ug
89-953-689 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-690 Supplier Abnova™ Supplier No. H00004686P02L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein promotes high-affinity mRNA-cap binding and associates with the CTD of RNA polymerase II. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. [provided by RefSeq]

Sequence: MSRRRHSDENDGGQPHKRRKTSDANETEDHLESLICKVGEKSACSLESNLEGLAGVLEADLPNYKSKILRLLCTVARLLPEKLTIYTTLVGLLNARNYNFGGEFVEAMIRQLKESLKANNYNEAVYLVRFLSDLVNCHVIAAPSMVAMFENFVSVTQEEDVPQVRRDWYVYAFLSSLPWVGKELYEKKDAEMDRIFANTESYLKRRQKTHVPMLQVWTADKPHPQEEYLDCLWAQIQKLKKDRWQERHILRPYLAFDSILCEALQHNLPPFTPPPHTEDSVYPMPRVIFRMFDYTDDPEGPVMPGSHSVERFVIEENLHCIIKSHWKERKTCAAQLVSYPGKNKIPLNYHIVEVIFAELFQLPAPPHIDVMYTTLLIELCKLQPGSLPQVLAQATEMLYMRLDTMNTTCVDRFINWFSHHLSNFQFRWSWEDWSDCLSQDPESPKPKFVREVLEKCMRLSYHQRILDIVPPTFSALCPANPTCIYKYGDESSNSLPGHSVALCLAVAFKSKATNDEIFSILKDVPNPNQDDDDDEGFSFNPLKIEVFVQTLLHLAAKSFSHSFSALAKFHEVFKTLAESDEGKLHVLRVMFEVWRNHPQMIAVLVDKMIRTQIVDCAAVANWIFSSELSRDFTRLFVWEILHSTIRKMNKHVLKIQKELEEAKEKLARQHKRRSDDDDRSSDRKDGVLEEQIERLQEKVESAQSEQKNLFLVIFQRFIMILTEHLVRCETDGTSVLTPWYKNCIERLQQIFLQHHQIIQQYMVTLENLLFTAELDPHILAVFQQFCALQA

Specifications

Accession Number NP_002477.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4686
Molecular Weight (g/mol) 118.2kDa
Name NCBP1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MSRRRHSDENDGGQPHKRRKTSDANETEDHLESLICKVGEKSACSLESNLEGLAGVLEADLPNYKSKILRLLCTVARLLPEKLTIYTTLVGLLNARNYNFGGEFVEAMIRQLKESLKANNYNEAVYLVRFLSDLVNCHVIAAPSMVAMFENFVSVTQEEDVPQVRRDWYVYAFLSSLPWVGKELYEKKDAEMDRIFANTESYLKRRQKTHVPMLQVWTADKPHPQEEYLDCLWAQIQKLKKDRWQERHILRPYLAFDSILCEALQHNLPPFTPPPHTEDSVYPMPRVIFRMFDYTDDPEGPVMPGSHSVERFVIEENLHCIIKSHWKERKTCAAQLVSYPGKNKIPLNYHIVEVIFAELFQLPAPPHIDVMYTTLLIELCKLQPGSLPQVLAQATEMLYMRLDTMNTTCVDRFINWFSHHLSNFQFRWSWEDWSDCLSQDPESPKPKFVREVLEKCMRLSYHQRILDIVPPTFSALCPANPTCIYKYGDESSNSLPGHSVALCLAVAFKSKATNDEIFSILKDVPNPNQDDDDDEGFSFNPLKIEVFVQTLLHLAAKSFSHSFSALAKFHEVFKTLAESDEGKLHVLRVMFEVWRNHPQMIAVLVDKMIRTQIVDCAAVANWIFSSELSRDFTRLFVWEILHSTIRKMNKHVLKIQKELEEAKEKLARQHKRRSDDDDRSSDRKDGVLEEQIERLQEKVESAQSEQKNLFLVIFQRFIMILTEHLVRCETDGTSVLTPWYKNCIERLQQIFLQHHQIIQQYMVTLENLLFTAELDPHILAVFQQFCALQA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CBP80/MGC2087/NCBP
Common Name NCBP1
Gene Symbol NCBP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.