Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCKIPSD Full-length ORF (NP_909119.1, 1 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-725 25 μg
89-953-724 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-725 Supplier Abnova™ Supplier No. H00051517P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintenance of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Sequence: MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHHRKETLSRRGPSASSVAVMTSSTSDHHLDAAAARQPNGVCRAGFERQHSLPSSEHLGADGGLYQIPPQPRRAAPTTPPPPVKRRDREALMASGSGGHNTMPSGGNSVSSGSSVSSTSLDTLYTSSSPSEPGSSCSPTPPPVPRRGTHTTVSQVQPPPSKASAPEPPAEEEVATGTTSASDDLEALGTLSLGTTEEKAAAEAAVPRTIGAELMELVRRNTGLSHELCRVAIGIIVGHIQASVPASSPVMEQVLLSLVEGKDLSMALPSGQVCHDQQRLEVIFADLARRKDDAQQRSWALYEDEGVIRCYLEELLHILTDADPEVCKKMCKRNEFESVLALVAYYQMEHRASLRLLLLKCFGAMCSLDAAIISTLVSSVLPVELARDMQTDTQDHQKLCYSALILAMVFSMGEAVPYAHYEHLGTPFAQFLLNIVEDGLPLDTTEQLPDLCVNLLLALNLHLPAADQNVIMAALSKHANVKIFSEKLLLLLNRGDDPVRIFKHEPQPPHSVLKFLQDVFGSPATAAIFYHTDMMALIDITVRHIADLSPGDKLRMEYLSLMHAIVRTTPYLQHRHRLPDLQAILRRILNEEETSPQCQMDRMIVREMCKEFLVLGEAPS

Specifications

Accession Number NP_909119.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51517
Molecular Weight (g/mol) 104.6kDa
Name NCKIPSD (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHHRKETLSRRGPSASSVAVMTSSTSDHHLDAAAARQPNGVCRAGFERQHSLPSSEHLGADGGLYQIPPQPRRAAPTTPPPPVKRRDREALMASGSGGHNTMPSGGNSVSSGSSVSSTSLDTLYTSSSPSEPGSSCSPTPPPVPRRGTHTTVSQVQPPPSKASAPEPPAEEEVATGTTSASDDLEALGTLSLGTTEEKAAAEAAVPRTIGAELMELVRRNTGLSHELCRVAIGIIVGHIQASVPASSPVMEQVLLSLVEGKDLSMALPSGQVCHDQQRLEVIFADLARRKDDAQQRSWALYEDEGVIRCYLEELLHILTDADPEVCKKMCKRNEFESVLALVAYYQMEHRASLRLLLLKCFGAMCSLDAAIISTLVSSVLPVELARDMQTDTQDHQKLCYSALILAMVFSMGEAVPYAHYEHLGTPFAQFLLNIVEDGLPLDTTEQLPDLCVNLLLALNLHLPAADQNVIMAALSKHANVKIFSEKLLLLLNRGDDPVRIFKHEPQPPHSVLKFLQDVFGSPATAAIFYHTDMMALIDITVRHIADLSPGDKLRMEYLSLMHAIVRTTPYLQHRHRLPDLQAILRRILNEEETSPQCQMDRMIVREMCKEFLVLGEAPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AF3P21/DIP/DIP1/MGC23891/ORF1/SPIN90/WASLBP/WISH
Common Name NCKIPSD
Gene Symbol NCKIPSD
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.