Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NDRG2 Full-length ORF (AAH11240.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953788 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-788 10 μg
89-953-789 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-788 Supplier Abnova™ Supplier No. H00057447P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MAELQEVQITEEKPLLPGQTPEAAKEAELAARILLDQGQTHSVETPYVSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPVFPLGYQYPSLDQLADMIPCVLQYLNFSTIIGVGVGAGAYILARYALNHPDTVEGLVLINIDPNAKGWMDWAAHKLTGLTSSIPEMILGHLFSQEELSGNSELIQKYRNIITHAPNLDNIELYWNSYNNRRDLNFERGGDITLRCPVMLVVECNSKLDPTQTSFLKMADSGGQPQLTQPGKLTEAFKYFLQGMGYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPGHTMEVSC

Specifications

Accession Number AAH11240.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57447
Molecular Weight (g/mol) 66.1kDa
Name NDRG2 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAELQEVQITEEKPLLPGQTPEAAKEAELAARILLDQGQTHSVETPYVSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPVFPLGYQYPSLDQLADMIPCVLQYLNFSTIIGVGVGAGAYILARYALNHPDTVEGLVLINIDPNAKGWMDWAAHKLTGLTSSIPEMILGHLFSQEELSGNSELIQKYRNIITHAPNLDNIELYWNSYNNRRDLNFERGGDITLRCPVMLVVECNSKLDPTQTSFLKMADSGGQPQLTQPGKLTEAFKYFLQGMGYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPGHTMEVSC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781G1938/FLJ25522/KIAA1248/SYLD
Common Name NDRG2
Gene Symbol NDRG2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.