Learn More
Abnova™ Human NDRG2 Full-length ORF (AAH11240.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH11240.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 57447 |
| Molecular Weight (g/mol) | 66.1kDa |
| Name | NDRG2 (Human) Recombinant Protein (P02) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MAELQEVQITEEKPLLPGQTPEAAKEAELAARILLDQGQTHSVETPYVSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPVFPLGYQYPSLDQLADMIPCVLQYLNFSTIIGVGVGAGAYILARYALNHPDTVEGLVLINIDPNAKGWMDWAAHKLTGLTSSIPEMILGHLFSQEELSGNSELIQKYRNIITHAPNLDNIELYWNSYNNRRDLNFERGGDITLRCPVMLVVECNSKLDPTQTSFLKMADSGGQPQLTQPGKLTEAFKYFLQGMGYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPGHTMEVSC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.