Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NEDD9/CASL Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10006429
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10006429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10006429 Supplier R&D Systems Supplier No. AF5459N100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

NEDD9/CASL/HEF1 Polyclonal antibody specifically detects NEDD9/CASL/HEF1 in Human samples. It is validated for Western Blot

Specifications

Antigen NEDD9/CASL/HEF1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias Cas scaffolding protein family member 2, CAS2, CASL, CAS-LCASS2cas-like docking, CASLdJ49G10.2, CRK-associated substrate-related protein, dJ49G10.2 (Enhancer of Filamentation 1 (HEF1)), dJ761I2.1, dJ761I2.1 (enhancer of filamentation (HEF1)), enhancer of filamentation 1, HEF1, HEF1Crk-associated substrate related, NEDD-9, Neural precursor cell expressed developmentally down-regulated protein 9, neural precursor cell expressed, developmentally down-regulated 9, p105, Renal carcinoma antigen NY-REN-12
Host Species Goat
Immunogen E. coli-derived recombinant human NEDD9, Lys153-Asn366, Accession # Q14511
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4739
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.