Learn More
Invitrogen™ Human NELL-1 ELISA Kit
Description
Human NELL-1 quantitates human NELL-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NELL-1.
Specifications
Specifications
| Accession Number | B230343H07Rik; IDH3GL; l7R6; Nell1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1 |
| Assay Range | 10 to 200 ng/mL |
| Assay Sensitivity | 10 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 4745 |
| Gene Symbol | NELL1 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.