Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Nephrin DuoSet ELISA
SDP

Catalog No. DY426905 Shop All Novus Products
Change view
Click to view available options
Sufficient For:
Five 96-well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DY426905 Five 96-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY426905 Supplier R&D Systems™ Supplier No. DY426905

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For determination of natural and recombinant human nephrin protein.

The Human Nephrin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Nephrin.

Specifications

Assay Range 0.37 to 90pg/mL
Immunoassay Kit Format 96-well Plate
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For Five 96-well plates
Target Species Human
Gene ID (Entrez) 4868 (Human); 54631 (Mouse); 64563 (Rat)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target Interferon gamma
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.