Learn More
Invitrogen™ Human Nestin/MMEL1 ELISA Kit
Description
Human Nestin/MMEL1 quantitates human Nestin/MMEL1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nestin/MMEL1.
Specifications
Specifications
| Accession Number | AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; nestin |
| Assay Range | 0.32 to 75 ng/mL |
| Assay Sensitivity | 0.32 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 10763 |
| Gene Symbol | Nes |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.