Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NETO1 Partial ORF (NP_620416.1, 434 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-002 25 μg
89-954-001 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-002 Supplier Abnova™ Supplier No. H00081832Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants encoding distinct proteins have been described. [provided by RefSeq]

Sequence: GSQLSSTKGSRSNLSTRDASILTEMPTQPGKPLIPPMNRRNILVMKHNYSQDAADACDIDEIEEVPTTSHRLSRHDKAVQRFCLIGSLSKHESEYNTTRV

Specifications

Accession Number NP_620416.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 81832
Molecular Weight (g/mol) 36.74kDa
Name NETO1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen GSQLSSTKGSRSNLSTRDASILTEMPTQPGKPLIPPMNRRNILVMKHNYSQDAADACDIDEIEEVPTTSHRLSRHDKAVQRFCLIGSLSKHESEYNTTRV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BCTL1/BTCL1
Common Name NETO1
Gene Symbol NETO1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.