Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neuropeptide Y/NPY Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10007731
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10007731 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10007731 Supplier R&D Systems Supplier No. FAB85171V100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Neuropeptide Y Monoclonal antibody specifically detects Neuropeptide Y in Human samples. It is validated for ELISA

Specifications

Antigen Neuropeptide Y
Applications ELISA
Classification Monoclonal
Clone 904022
Conjugate Alexa Fluor 405
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, NPY, PYY4, seprase
Host Species Mouse
Immunogen Human Neuropeptide Y/NPY conjugated to KLH, Accession # P01303
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4852
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.