Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Neuropeptide Y/NPY DuoSet ELISA
SDP

Catalog No. DY851705 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY851705 1 Kit (5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY851705 Supplier R&D Systems™ Supplier No. DY851705

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Human Neuropeptide Y/NPY DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Neuropeptide Y.

The Human Neuropeptide Y/NPY DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Neuropeptide Y.

Specifications

Assay Range 312.00 - 10000pg/mL
Product Line R&D Systems
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4852
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.