Learn More
Description
- NTM neurotrimin [ Homo sapiens (human) ]
- Also known as: HNT; NTRI; CEPU1; IGLON2
- The Neurotrimin/HNT insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- Neurotrimin is a glycosylphosphatidylinositol (GPI)-anchored cell adhesion molecule expressed on neuronal population.
- NTM is a member of the IgLON (LAMP, OBCAM, Ntm) family of immunoglobulin (Ig) domaincontaining glycosylphosphatidylinositol (GPI)- anchored cell adhesion molecules.
- It may promote neurite outgrowth and adhesion via a homophilic mechanism.
- Alternatively spliced transcripts encoding different proteins have been described.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hNTM |
| Accession Number | NP_057606 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1048 bp |
| Gene ID (Entrez) | 50863 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
