Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Neurturin Recombinant Protein, PeproTech®

Catalog No. 450115UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-11-5UG 5 μg
45011100UG 100 μg
45-011-01M 2 x 500 μg
45011250UG 250 μg
45011500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 450-11-5UG Supplier Gibco™ Supplier No. 450115UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

450-11-1MG will be provided as 2 x 500 μg (450-11-500UG). The functional form of human neurturin is a disulfide-linked homodimer, of two 11.8 kDa polypeptide monomers (204 total amino acid residues). Each monomer contains seven conserved cysteine residues, one of which (Cys 69) is used for inter-chain disulfide bridging, and the others are involved in the intramolecular ring formation known as the cysteine-knot configuration. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99748
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4902
Molecular Weight (g/mol) 23.6 kDa
Name Human Neurturin
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias neurturin; NRTN; NTN; prepro-neurturin
Common Name Neurturin
Gene Symbol NRTN
Biological Activity Human Neurturin at a concentration of 100 ng/ml will support the survival of 65% of newborn rat sympathetic neurons.
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.