Learn More
Specifications
Specifications
| Assay Range | 125.00 - 8000 pg/mL |
| Detection Method | Sandwich ELISA |
| Target Species | Human |
| Gene ID (Entrez) | 4747 |
| Quantity | 1 Kit (for 5 Plates) |
| Synonym | 68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
