Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human NF-M ELISA Kit (Colorimetric)
SDP

Catalog No. NB001544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001544 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001544 Supplier Novus Biologicals™ Supplier No. NBP278758
Only null left
Add to Cart
Add to Cart

Specifications

Accession Number P07197
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human NEF3 in samples. No significant cross-reactivity or interference between Human NEF3 and analogues was observed
For Use With (Application) ELISA
Gene ID (Entrez) 4741
Gene Symbol NEFM
Host Species Human
Quantity 1 Kit
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target NF-M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.