Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NFE2L3 Full-length ORF (NP_004280.4, 1 a.a. - 694 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954074 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-074 25 μg
89-954-073 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-074 Supplier Abnova™ Supplier No. H00009603P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the cap 'n' collar basic-region leucine zipper family of transcription factors. The encoded protein heterodimerizes with small musculoaponeurotic fibrosarcoma factors to bind antioxidant response elements in target genes. This protein is a membrane bound glycoprotein that is targeted to the endoplasmic reticulum and the nuclear envelope. Pseudogenes of this gene are found on chromosomes 16, 17, and 18. [provided by RefSeq]

Sequence: MKHLKRWWSAGGGLLHLTLLLSLAGLRVDLDLYLLLPPPTLLQDELLFLGGPASSAYALSPFSASGGWGRAGHLHPKGRELDPAAPPEGQLLREVRALGVPFVPRTSVDAWLVHSVAAGSADEAHGLLGAAAASSTGGAGASVDGGSQAVQGGGGDPRAARSGPLDAGEEEKAPAEPTAQVPDAGGCASEENGVLREKHEAVDHSSQHEENEERVSAQKENSLQQNDDDENKIAEKPDWEAEKTTESRNERHLNGTDTSFSLEDLFQLLSSQPENSLEGISLGDILLPGSISDGMNSSAHYHVNFSQAISQDVNLHEAILLCPNNTFRRDPTARTSQSQEPFLQLNSHTTNPEQTLPGTNLTGFLSPVDNHMRNLTSQDLLYDLDINIFDEINLMSLATEDNFDPIDVSQLFDEPDSDSGLSLDSSHNNTSVIKSNSSHSVCDEGAIGYCTDHESSSHHDLEGAVGGYYPEPSKLCHLDQSDSDFHGDLTFQHVFHNHTYHLQPTAPESTSEPFPWPGKSQKIRSRYLEDTDRNLSRDEQRAKALHIPFSVDEIVGMPVDSFNSMLSRYYLTDLQVSLIRDIRRRGKNKVAAQNCRKRKLDIILNLEDDVCNLQAKKETLKREQAQCNKAINIMKQKLHDLYHDIFSRLRDDQGRPVNPNHYALQCTHDGSILIVPKELVASGHKKETQKGKRK

Specifications

Accession Number NP_004280.4
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9603
Molecular Weight (g/mol) 102.6kDa
Name NFE2L3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKHLKRWWSAGGGLLHLTLLLSLAGLRVDLDLYLLLPPPTLLQDELLFLGGPASSAYALSPFSASGGWGRAGHLHPKGRELDPAAPPEGQLLREVRALGVPFVPRTSVDAWLVHSVAAGSADEAHGLLGAAAASSTGGAGASVDGGSQAVQGGGGDPRAARSGPLDAGEEEKAPAEPTAQVPDAGGCASEENGVLREKHEAVDHSSQHEENEERVSAQKENSLQQNDDDENKIAEKPDWEAEKTTESRNERHLNGTDTSFSLEDLFQLLSSQPENSLEGISLGDILLPGSISDGMNSSAHYHVNFSQAISQDVNLHEAILLCPNNTFRRDPTARTSQSQEPFLQLNSHTTNPEQTLPGTNLTGFLSPVDNHMRNLTSQDLLYDLDINIFDEINLMSLATEDNFDPIDVSQLFDEPDSDSGLSLDSSHNNTSVIKSNSSHSVCDEGAIGYCTDHESSSHHDLEGAVGGYYPEPSKLCHLDQSDSDFHGDLTFQHVFHNHTYHLQPTAPESTSEPFPWPGKSQKIRSRYLEDTDRNLSRDEQRAKALHIPFSVDEIVGMPVDSFNSMLSRYYLTDLQVSLIRDIRRRGKNKVAAQNCRKRKLDIILNLEDDVCNLQAKKETLKREQAQCNKAINIMKQKLHDLYHDIFSRLRDDQGRPVNPNHYALQCTHDGSILIVPKELVASGHKKETQKGKRK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NRF3
Common Name NFE2L3
Gene Symbol NFE2L3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.