Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NFKBIZ Full-length ORF (NP_113607.1, 1 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954106 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-106 25 μg
89-954-105 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-106 Supplier Abnova™ Supplier No. H00064332P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the ankyrin-repeat family and is induced by lipopolysaccharide (LPS). The C-terminal portion of the encoded product which contains the ankyrin repeats, shares high sequence similarity with the I kappa B family of proteins. The latter are known to play a role in inflammatory responses to LPS by their interaction with NF-B proteins through ankyrin-repeat domains. Studies in mouse indicate that this gene product is one of the nuclear I kappa B proteins and an activator of IL-6 production. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPY

Specifications

Accession Number NP_113607.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 64332
Molecular Weight (g/mol) 104.5kDa
Name NFKBIZ (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ30225/FLJ34463/IKBZ/INAP/MAIL
Common Name NFKBIZ
Gene Symbol NFKBIZ
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.