Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NFYC Full-length ORF (NP_055038.2, 1 a.a. - 335 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954122 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-122 25 ug
89-954-121 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-122 Supplier Abnova™ Supplier No. H00004802P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes one subunit of a trimeric complex forming a highly conserved transcription factor that binds with high specificity to CCAAT motifs in the promoters of a variety of genes. The encoded protein, subunit C, forms a tight dimer with the B subunit, a prerequisite for subunit A association. The resulting trimer binds to DNA with high specificity and affinity. Subunits B and C each contain a histone-like motif. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MSTEGGFGGTSSSDAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFLIDIVPRDELKPPKRQEEVRQSVTPAEPVQYYFTLAQQPTAVQVQGQQQGQQTTSSTTTIQPGQIIIAQPQQGQTTPVTMQVGEGQQVQIVQAQPQGQAQQAQSGTGQTMQVMQQIITNTGEIQQIPVQLNAGQLQYIRLAQPVSGTQVVQGQIQTLATNAQQITQTEVQQGQQQFSQFTDGQQLYQIQQVTMPAGQDLAQPMFIQSANQPSDGQAPQVTGD

Specifications

Accession Number NP_055038.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4802
Molecular Weight (g/mol) 63.6kDa
Name NFYC (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MSTEGGFGGTSSSDAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFLIDIVPRDELKPPKRQEEVRQSVTPAEPVQYYFTLAQQPTAVQVQGQQQGQQTTSSTTTIQPGQIIIAQPQQGQTTPVTMQVGEGQQVQIVQAQPQGQAQQAQSGTGQTMQVMQQIITNTGEIQQIPVQLNAGQLQYIRLAQPVSGTQVVQGQIQTLATNAQQITQTEVQQGQQQFSQFTDGQQLYQIQQVTMPAGQDLAQPMFIQSANQPSDGQAPQVTGD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CBF-C/CBFC/DKFZp667G242/FLJ45775/H1TF2A/HAP5/HSM/NF-YC
Common Name NFYC
Gene Symbol NFYC
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.