Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NGLY1 Full-length ORF (NP_060767.2, 1 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-141 10 μg
89-954-142 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-141 Supplier Abnova™ Supplier No. H00055768P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MAAAALGSSSGSASPAVAELCQNTPETFLEASKLLLTYADNILRNPNDEKYRSIRIGNTAFSTRLLPVRGAVECLFEMGFEEGETHLIFPKKASVEQLQKIRDLIAIERSSRLDGSNKSHKVKSSQQPAASTQLPTTPSSNPSGLNQHTRNRQGQSSDPPSASTVAADSAILEVLQSNIQHVLVYENPALQEKALACIPVQELKRKSQEKLSRARKLDKGINISDEDFLLLELLHWFKEEFFHWVNNVLCSKCGGQTRSRDRSLLPSDDELKWGAKEVEDHYCDACQFSNRFPRYNNPEKLLETRCGRCGEWANCFTLCCRAVGFEARYVWDYTDHVWTEVYSPSQQRWLHCDACEDVCDKPLLYEIGWGKKLSYVIAFSKDEVVDVTWRYSCKHEEVIARRTKVKEALLRDTINGLNKQRQLFLSENRRKELLQRIIVELVEFISPKTPKPGELGGRISGSVAWRVARGEMGLQRKETLFIPCENEKISKQLHLCYNIVKDRYVRVSNNNQTISGWENGVWKMESIFRKVETDWHMVYLARKEGSSFAYISWKFECGSVGLKVDSISIRTSSQTFQTGTVEWKLRSDTAQVELTGDNSLHSYADFSGATEVILEAELSRGDGDVAWQHTQLFRQSLNDHEENCLEIIIKFSDL

Specifications

Accession Number NP_060767.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55768
Molecular Weight (g/mol) 100.8kDa
Name NGLY1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAAAALGSSSGSASPAVAELCQNTPETFLEASKLLLTYADNILRNPNDEKYRSIRIGNTAFSTRLLPVRGAVECLFEMGFEEGETHLIFPKKASVEQLQKIRDLIAIERSSRLDGSNKSHKVKSSQQPAASTQLPTTPSSNPSGLNQHTRNRQGQSSDPPSASTVAADSAILEVLQSNIQHVLVYENPALQEKALACIPVQELKRKSQEKLSRARKLDKGINISDEDFLLLELLHWFKEEFFHWVNNVLCSKCGGQTRSRDRSLLPSDDELKWGAKEVEDHYCDACQFSNRFPRYNNPEKLLETRCGRCGEWANCFTLCCRAVGFEARYVWDYTDHVWTEVYSPSQQRWLHCDACEDVCDKPLLYEIGWGKKLSYVIAFSKDEVVDVTWRYSCKHEEVIARRTKVKEALLRDTINGLNKQRQLFLSENRRKELLQRIIVELVEFISPKTPKPGELGGRISGSVAWRVARGEMGLQRKETLFIPCENEKISKQLHLCYNIVKDRYVRVSNNNQTISGWENGVWKMESIFRKVETDWHMVYLARKEGSSFAYISWKFECGSVGLKVDSISIRTSSQTFQTGTVEWKLRSDTAQVELTGDNSLHSYADFSGATEVILEAELSRGDGDVAWQHTQLFRQSLNDHEENCLEIIIKFSDL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ11005/FLJ12409/PNG1
Common Name NGLY1
Gene Symbol NGLY1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.