Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NID2 Full-length ORF (AAH35608.1, 1 a.a. - 969 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954176 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-176 10 μg
89-954-177 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-176 Supplier Abnova™ Supplier No. H00022795P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Basement membranes, which are composed of type IV collagens (see MIM 120130), laminins (see LAMC1; MIM 150290), perlecan (HSPG2; MIM 142461), and nidogen (see NID1; MIM 131390), are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.[supplied by OMIM]

Sequence: MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK

Specifications

Accession Number AAH35608.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22795
Molecular Weight (g/mol) 133.5kDa
Name NID2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name NID2
Gene Symbol NID2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.