Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human NIPP1 (NP_054829.2) VersaClone cDNA
SDP

Catalog No. RDC3800 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3800 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC3800 Supplier R&D Systems™ Supplier No. RDC3800

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Gene NIPP1
Accession Number NP_054829.2
Gene ID (Entrez) 5511
Gene Alias activator of RNA decay, ard-1, ARD1, ARD1nuclear inhibitor of protein phosphatase 1, NIPP1nuclear inhibitor of protein phosphatase-1, NIPP-1nuclear subunit of PP-1, PPP1R8, PRO2047, Protein phosphatase 1 regulatory inhibitor subunit 8, protein phosphatase 1 regulatory subunit 8, protein phosphatase 1, regulatory (inhibitor) subunit 8, RNase E
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.