Learn More
Sartorius Human NK Cell Companion Kit
Supplier:  Sartorius 97089
Encompass
The Intellicyt Human NK Cell Companion Kits can be used to measure additional cytokines besides those already included in the Human NK Cell Mediated Killing Kit.
Catalog No. 50-289-1738