Learn More
Description
- KLRC1 killer cell lectin like receptor C1 [ Homo sapiens (human) ]
- Also known as: NKG2; NKG2A; CD159A
- NKG2A and NKG2C are type II transmembrane proteins with extracellular C-type lectin domains.
- These proteins heterodimerize with CD94 to form complexes that recognize HLA-E as ligand.
- The NKG2A/CD94 complex delivers an inhibitory signal resulting from ITIMs within the NKG2A cytoplasmic domain.
- NKG2C/CD94 provides an activating signal due to association with the ITAM-containing DAP12 adapter protein.
- These complexes are expressed on NK and T cell subsets.
- The NKG2A/CD159a/KLRC1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hKLRC1 |
| Accession Number | NP_002250 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 715 bp |
| Gene ID (Entrez) | 3821 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
