Learn More
Description
- KLRC3 killer cell lectin like receptor C3 [ Homo sapiens (human) ]
- Also known as: NKG2E; NKG2-E
- NKG2H belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells.
- This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain.
- Alternatively spliced transcripts encoding different proteins have been described.
- The NKG2H insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hKLRC3 |
| Accession Number | NP_002252 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 736 bp |
| Gene ID (Entrez) | 3823 |
| Gene Alias | KLRC3, NKG2E, NKG2-E |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
