Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NKp44/NCR2 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4716532
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB22491A02 25 Tests
FAB22491A10 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB22491A02 Supplier R&D Systems Supplier No. FAB22491A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

NKp44/NCR2 Monoclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NKp44/NCR2
Applications Flow Cytometry
Classification Monoclonal
Clone 253415
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. CAB52289
Gene Alias CD336, CD336 antigen, LY95, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NCR2, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1
Gene Symbols NCR2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2 Gln22-Pro190 Accession # CAB52289
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9436
Test Specificity Detects human NKp44/NCR2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp46, rhNKp80, or recombinant mouse NKp46 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.