Learn More
ACROBiosystems Human NKp46 Protein Fc Tag
Supplier:  ACROBiosystems NC1H52591MG
Encompass_Preferred
Human NKp46 Protein Fc Tag (NC1-H5259) is expressed from human 293 cells (HEK293). It contains AA His 129 - Thr 211 (Accession # O76036-1).
Catalog No. 50-253-4401
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.