Learn More
Biomatik Corporation Human NLR Family, Pyrin Domain Containing Protein 7 (NLRP7) ELISA Kit
Supplier:  Biomatik Corporation EKU11316
Encompass_Preferred
Human NLR Family, Pyrin Domain Containing Protein 7 (NLRP7) ELISA Kit for detecting NLRP7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WX94. Gene ID: 199713. Target Synonyms: NALP7; PYPAF3; NOD12; PAN7; CLR19.4; Nucleotide-Binding Oligomerization Domain,Leucine Rich Repeat And Pyrin Domain Containing 7; PYRIN-containing APAF1-like 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8169