Learn More
Novus Biologicals™ Human NLRP3/NALP3 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP318216
This item is not returnable.
View return policy
Specifications
0.31-20 ng/ml | |
ELISA Kit (Colorimetric) | |
ELISA | |
NLRP3 | |
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 |
0.16 ng/ml | |
Biological Fluids, Cell Culture Supernatant, Cell Lysates, Plasma, Serum, Tissue Homogenates | |
114548 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |