Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human NNT-1/BCSF-3 Recombinant Protein, PeproTech®

Catalog No. 45018100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45018100UG 100 μg
450-18-10UG 10 μg
450-18-1MG 1 mg
45018250UG 250 μg
450-18-2UG 2 μg
45018500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45018100UG Supplier Gibco™ Supplier No. 45018100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human NNT-1/BCSF-3 is a 22.4 kDa protein containing 199 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

NNT-1/BCSF-3 is a neurotrophic factor with B-cell-stimulating capabilities. Expressed in the lymph nodes and spleen, NNT-1/BCSF-3 activates glycoprotein 130 (gp130) and leukemia inhibitory factor receptor member b (LIFR-b) by binding and inducing the tyrosine phosphorylation of these receptors. In vitro, it supports the survival of chick embryo motor and sympathetic neurons. In mice, NNT-1/BCSF-3 induces serum amyloid A, causes body weight loss, and B cell hyperplasia associated with increases in serum IgG and IgM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBD9
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 23529
Molecular Weight (g/mol) 22.4 kDa
Name Human NNT-1/BCSF-3
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias b cell-stimulating factor 3; B-cell stimulating factor 3; B-cell stimulatory factor 3; B-cell-stimulating factor 3; Bsf3; BSF-3; Cardiotrophin-like cytokine factor 1; CISS2; Clc; CLCF1; CRLF1 associated cytokine-like factor 1; neurotrophin-1/B-cell stimulating factor-3; NNT1; NNT-1; NNT-1/BSF-3; Novel neurotrophin-1; NR6
Common Name NNT1
Gene Symbol CLCF1
Biological Activity NT-1/BCSF-3 weakly supports chick E8 DRG neurite outgrowth (at a concentration of 1.0 ng/ml)
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.