Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NOC2L Full-length ORF (AAH03555.1, 1 a.a. - 749 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954343 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-343 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-954-343 Supplier Abnova™ Supplier No. H00026155P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Histone modification by histone acetyltransferases (HAT) and histone deacetylases (HDAC) can control major aspects of transcriptional regulation. NOC2L represents a novel HDAC-independent inhibitor of histone acetyltransferase (INHAT) (Hublitz et al., 2005 [PubMed 16322561]).[supplied by OMIM]

Sequence: MAAAGSRKRRLAELTVDEFLASGFDSESESESENSPQAETREAREAARSPDKPGGSPSASRRKGRASEHKDQLSRLKDRDPEFYKFLQENDQSLLNFSDSDSSEEEEGPFHSLPDVLEEASEEEDGAEEGEDGDRVPRGLKGKKNSVPVTVAMVERWKQAAKQRLTPKLFHEVVQAFRAAVATTRGDQESAEANKFQVTDSAAFNALVTFCIRDLIGCLQKLLFGKVAKDSSRMLQPSSSPLWGKLRVDIKAYLGSAIQLVSCLSETTVLAAVLRHISVLVPCFLTFPKQCRMLLKRMVVVWSTGEESLRVLAFLVLSRVCRHKKDTFLGPVLKQMYITYVRNCKFTSPGALPFISFMQWTLTELLALEPGVAYQHAFLYIRQLAIHLRNAMTTRKKETYQSVYNWQYVHCLFLWCRVLSTAGPSEALQPLVYPLAQVIIGCIKLIPTARFYPLRMHCIRALTLLSGSSGAFIPVLPFILEMFQQVDFNRKPGRMSSKPINFSVILKLSNVNLQEKAYRDGLVEQLYDLTLEYLHSQAHCIGFPELVLPVVLQLKSFLRECKVANYCRQVQQLLGKVQENSAYICSRRQRVSFGVSEQQAVEAWEKLTREEGTPLTLYYSHWRKLRDREIQLEISGKERLEDLNFPEIKRRKMADRKDEDRKQFKDLFDLNSSEEDDTEGFSERGILRPLSTRHGVEDDEEDEEEGEEDSSNSEDGDPDAEAGLAPGELQQLAQGPEDELEDLQLSEDD

Specifications

Accession Number AAH03555.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26155
Molecular Weight (g/mol) 111.3kDa
Name NOC2L (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAAAGSRKRRLAELTVDEFLASGFDSESESESENSPQAETREAREAARSPDKPGGSPSASRRKGRASEHKDQLSRLKDRDPEFYKFLQENDQSLLNFSDSDSSEEEEGPFHSLPDVLEEASEEEDGAEEGEDGDRVPRGLKGKKNSVPVTVAMVERWKQAAKQRLTPKLFHEVVQAFRAAVATTRGDQESAEANKFQVTDSAAFNALVTFCIRDLIGCLQKLLFGKVAKDSSRMLQPSSSPLWGKLRVDIKAYLGSAIQLVSCLSETTVLAAVLRHISVLVPCFLTFPKQCRMLLKRMVVVWSTGEESLRVLAFLVLSRVCRHKKDTFLGPVLKQMYITYVRNCKFTSPGALPFISFMQWTLTELLALEPGVAYQHAFLYIRQLAIHLRNAMTTRKKETYQSVYNWQYVHCLFLWCRVLSTAGPSEALQPLVYPLAQVIIGCIKLIPTARFYPLRMHCIRALTLLSGSSGAFIPVLPFILEMFQQVDFNRKPGRMSSKPINFSVILKLSNVNLQEKAYRDGLVEQLYDLTLEYLHSQAHCIGFPELVLPVVLQLKSFLRECKVANYCRQVQQLLGKVQENSAYICSRRQRVSFGVSEQQAVEAWEKLTREEGTPLTLYYSHWRKLRDREIQLEISGKERLEDLNFPEIKRRKMADRKDEDRKQFKDLFDLNSSEEDDTEGFSERGILRPLSTRHGVEDDEEDEEEGEEDSSNSEDGDPDAEAGLAPGELQQLAQGPEDELEDLQLSEDD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp564C186/FLJ35172/NIR
Common Name NOC2L
Gene Symbol NOC2L
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.