Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Nogo-B/RTN 4B ELISA Kit

Catalog No. EH344RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH344RB 96 Tests
EH344RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH344RB Supplier Invitrogen™ Supplier No. EH344RB
Add to Cart
Add to Cart

ELISA

Human Nogo-B/RTN 4B quantitates human Nogo-B/RTN 4B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nogo-B/RTN 4B.

Nogo-B is a member of the reticulon-4 (RTN4) family of proteins, primarily localized to the endoplasmic reticulum and expressed in many tissues, especially vascular endothelial and smooth muscle cells. It plays a key role in regulating vascular remodeling, cell migration, and tissue repair by influencing cytoskeletal organization and signaling pathways. Unlike Nogo-A, which is known for inhibiting nerve regeneration, Nogo-B is more involved in maintaining vascular homeostasis and modulating inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQC3
Assay Range 13-3000 pg/mL
Assay Sensitivity 13 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; foocen; Glut4 vesicle 20 kDa protein; GLUT4 vesicle 20kDa protein; Human NogoA; KIAA0886; mKIAA0886; mKIAA4153; My043; My043 protein; Nbla00271; Nbla10545; neurite growth inhibitor 220; neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NgA; NI220/250; NI-220/250; NI-250; NOGO; nogo protein; Nogo-A; Nogo-B; NOGOC; Nogo-C; NSP; NSP-CL; r; rat N; rat NogoA; reticulon 4; reticulon 5; reticulon-4; Reticulon-5; Rtn4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; RTN-x; SP1507; Vp201110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; foocen; Glut4 vesicle 20 kDa protein; GLUT4 vesicle 20kDa protein; Human NogoA; KIAA0886; mKIAA0886; mKIAA4153; My043; My043 protein; Nbla00271; Nbla10545; neurite growth inhibitor 220; neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NgA; NI220/250; NI-220/250; NI-250; NOGO; nogo protein; Nogo-A; Nogo-B; NOGOC; Nogo-C; NSP; NSP-CL; r; rat N; rat NogoA; reticulon 4; reticulon 5; reticulon-4; Reticulon-5; Rtn4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; RTN-x; SP1507; Vp20;
Gene ID (Entrez) 57142
Gene Symbol Rtn4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Nogo-B
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.