Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Norrin ELISA Kit

Catalog No. EH345RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH345RB 96 Tests
EH345RBX5 5 x 96 Tests
EH345RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH345RB Supplier Invitrogen™ Supplier No. EH345RB
Add to Cart
Add to Cart

ELISA

Human Norrin quantitates human Norrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Norrin.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number EVR2; FEVR; ND; Ndp; NDP, norrin cystine knot growth factor; Ndph; Norrie disease (pseudoglioma); Norrie disease (pseudoglioma) (human); Norrie disease homolog; norrie disease protein; Norrie disease protein homolog; Norrin; RGD1563968; X-linked exudative vitreoretinopathy 2 protein
Assay Range 0.41 to 100 ng/mL
Assay Sensitivity 0.41 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4693
Gene Symbol NDP
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target NDP
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.