Learn More
Description
- Also known as: MOX1; NOH1; NOH-1; GP91-2
- NOX1 is a member of the NADPH oxidase family of enzymes responsible for the catalytic oneelectron transfer of oxygen to generate superoxide or hydrogen peroxide.
- It participates in the regulation of cellular pH and is blocked by zinc.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hNOX1bed |
| Accession Number | NP_008983 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1708 bp |
| Gene ID (Entrez) | 27035 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
