Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nox4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7158801
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8158SP 25 μg
MAB8158100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8158SP Supplier R&D Systems Supplier No. MAB8158SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nox4 Monoclonal specifically detects Nox4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Nox4
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 996715
Conjugate Unconjugated
Dilution Immunohistochemistry 1-25 μg/mL, Immunocytochemistry 5-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9NPH5
Gene Alias EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOX, RENOXEC 1.6.3.-
Host Species Mouse
Immunogen E. coli-derived recombinant human NOX4, Lys450-Ser578, Accession No. Q9NPH5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 50507.0
Test Specificity Detects human NOX4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.