Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NOX4 Partial ORF (NP_058627.1, 479 a.a. - 578 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954424 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-424 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-954-424 Supplier Abnova™ Supplier No. H00050507Q01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the NOX-family of enzymes that functions as the catalytic subunit the NADPH oxidase complex. The encoded protein is localized to non-phagocytic cells where it acts as an oxygen sensor and catalyzes the reduction of molecular oxygen to various reactive oxygen species (ROS). The ROS generated by this protein have been implicated in numerous biological functions including signal transduction, cell differentiation and tumor cell growth. A pseudogene has been identified on the other arm of chromosome 11. Alternative splicing results in multiple transcript variants

Sequence: LHNKFWQENRPDYVNIQLYLSQTDGIQKIIGEKYHALNSRLFIGRPRWKLLFDEIAKYNRGKTVGVFCCGPNSLSKTLHKLSNQNNSYGTRFEYNKESFS

Specifications

Accession Number NP_058627.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 50507
Molecular Weight (g/mol) 36.74kDa
Name NOX4 (Human) Recombinant Protein (Q01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen LHNKFWQENRPDYVNIQLYLSQTDGIQKIIGEKYHALNSRLFIGRPRWKLLFDEIAKYNRGKTVGVFCCGPNSLSKTLHKLSNQNNSYGTRFEYNKESFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KOX/KOX-1/RENOX
Common Name NOX4
Gene Symbol NOX4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.