Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NOXO1 Partial ORF (AAH15917, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-430 10 μg
89-954-431 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-430 Supplier Abnova™ Supplier No. H00124056Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). NOX organizers, such as NOXO1, target NOX activators (see NOXA1; MIM 611255) to NOX and also target NOX to different subcellular compartments (Opitz et al., 2007 [PubMed 17189823]).[supplied by OMIM]

Sequence: MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRR

Specifications

Accession Number AAH15917
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 124056
Molecular Weight (g/mol) 36.74kDa
Name NOXO1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC20258/P41NOX/P41NOXA/P41NOXB/P41NOXC/SH3PXD5/SNX28
Common Name NOXO1
Gene Symbol NOXO1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.