Learn More
Sino Biological NPC1/Niemann-Pick C1 protein Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 16499T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NPC1/Niemann-Pick C1 protein (rh NPC1/Niemann-Pick C1 protein; Catalog#16499-H32H; NP_000262.2; Arg372-Phe622). NPC1/Niemann-Pick C1 protein specific IgG was purified by Human NPC1/Niemann-Pick C1 protein affinity chromatography.
Catalog No. 50-255-4020
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.