Learn More
ACROBiosystems Human Nr4a1 Protein His Tag
Supplier:  ACROBiosystems NR1H51441MG
Encompass_Preferred
Human Nr4a1 Protein His Tag (NR1-H5144) is expressed from E. coli cells. It contains AA Ser 351 - Phe 598 (Accession # P22736-1).
Catalog No. 50-253-4357
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.