Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NRG2 Full-length ORF (AAI66615.1) Recombinant Protein without tag

Catalog No. 89954624 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-624 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-954-624 Supplier Abnova™ Supplier No. H00009542G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

Neuregulin 2 (NRG2) is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1 (NRG1), another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: MRQVCCSALPPPPLEKGRCSSYSDSSSSSSERSSSSSSSSSESGSSSRSSSNNSSISRPAAPPEPRPQQQPQPRSPAARRAAARSRAAAAGGMRRDPAPGFSMLLFGVSLACYSPSLKSVQDQAYKAPVVVEGKVQGLVPAGGSSSNSTREPPASGRVALVKVLDKWPLRSGGLQREQVISVGSCVPLERNQRYIFFLEPTEQPLVFKTAFAPLDTNGKNLKKEVGKILCTDCATRPKLKKMKSQTGQVGEKQSLKCEAAAGNPQPSYRWFKDGKELNRSRDIRIKYGNGRKNSRLQFNKVKVEDAGEYVCEAENILGKDTVRGRLYVNSVSTTLSSWSGHARKCNETAKSYCVNGGVCYYIEGINQLSCKCPVGYTGDRCQQFAMVNFSKHLGFELKEAEELYQKRVLTITGICVALLVVGIVCVVAYCKTKKQRKQMHNHLRQNMCPAHQNRSLANGPSHPRLDPEEIQMADYISKNVPATDHVIRRETETTFSGSHSCSPSHHCSTATPTSSHRHESHTWSLERSESLTSDSQSGIMLSSVGTSKCNSPACVEARARRAAAYNLEERRRATAPPYHDSVDSLRDSPHSERYVSALTTPARLSPVDFHYSLATQVPTFEITSPNSAHAVSLPPAAPISYRLAEQQPLLRHPAPPGPGPGPGPGPGPGADMQRSYDSYYYPAAGPGPRRGTCALGGSLGSLPASPFRIPEDDEYETTQECAPPPPPRPRARGASRRTSAGPRRWRRSRLNGLAAQRARAARDSLSLSSGSGGGSASASDDDADDADGALAAESTPFLGLRGAHDALRSDSPPLCPAADSRTYYSLDSHSTRASSRHSRGPPPRAKQDSAPL

Specifications

Accession Number AAI66615.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 9542
Molecular Weight (g/mol) 93.8kDa
Name NRG2 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MRQVCCSALPPPPLEKGRCSSYSDSSSSSSERSSSSSSSSSESGSSSRSSSNNSSISRPAAPPEPRPQQQPQPRSPAARRAAARSRAAAAGGMRRDPAPGFSMLLFGVSLACYSPSLKSVQDQAYKAPVVVEGKVQGLVPAGGSSSNSTREPPASGRVALVKVLDKWPLRSGGLQREQVISVGSCVPLERNQRYIFFLEPTEQPLVFKTAFAPLDTNGKNLKKEVGKILCTDCATRPKLKKMKSQTGQVGEKQSLKCEAAAGNPQPSYRWFKDGKELNRSRDIRIKYGNGRKNSRLQFNKVKVEDAGEYVCEAENILGKDTVRGRLYVNSVSTTLSSWSGHARKCNETAKSYCVNGGVCYYIEGINQLSCKCPVGYTGDRCQQFAMVNFSKHLGFELKEAEELYQKRVLTITGICVALLVVGIVCVVAYCKTKKQRKQMHNHLRQNMCPAHQNRSLANGPSHPRLDPEEIQMADYISKNVPATDHVIRRETETTFSGSHSCSPSHHCSTATPTSSHRHESHTWSLERSESLTSDSQSGIMLSSVGTSKCNSPACVEARARRAAAYNLEERRRATAPPYHDSVDSLRDSPHSERYVSALTTPARLSPVDFHYSLATQVPTFEITSPNSAHAVSLPPAAPISYRLAEQQPLLRHPAPPGPGPGPGPGPGPGADMQRSYDSYYYPAAGPGPRRGTCALGGSLGSLPASPFRIPEDDEYETTQECAPPPPPRPRARGASRRTSAGPRRWRRSRLNGLAAQRARAARDSLSLSSGSGGGSASASDDDADDADGALAAESTPFLGLRGAHDALRSDSPPLCPAADSRTYYSLDSHSTRASSRHSRGPPPRAKQDSAPL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Don-1/HRG2/NTAK
Common Name NRG2
Gene Symbol NRG2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.