Learn More
Abnova™ Human NT5DC2 Full-length ORF (NP_075059.1, 1 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYR
Specifications
Specifications
| Accession Number | NP_075059.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 64943 |
| Name | NT5DC2 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 25 μg |
| Immunogen | MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYRGLQPVPDEEVIELYGGTQHIPLYQMSGFYGKGPSIKQFMDIFSLPEMALLSCVVDYFLGHSLEFDQAHLYKDVTDAIRDVHVKGLMYQWIEQDMEKYILRGDETFAVLSRLVAHGKQLFLITNSPFSFVDKGMRHMVGPDWRQLFDVVIVQADKPSFFTDRRKPFRKLDEKGSLQWDRITRLEKGKIYRQGNLFDFLRLTEWRGPRVLYFGDHLYSDLADLMLRHGWRTGAIIPELEREIRIINTEQYMHSLTWQQALTGLLERMQTYQDAESRQVLAAWMKERQELRCITKALFNAQFGSIFRTFHNPTYFSRRLVRFSDLYMASLSCLLNYRVDFTFYPRRTPLQHEAPLWMDQLCTGCMKTPFLGDMAHIR |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.