Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NTB-A/SLAMF6 ELISA Kit

Catalog No. EH350RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH350RB 96 Tests
EH350RBX5 5 x 96 Tests
EH350RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH350RB Supplier Invitrogen™ Supplier No. EH350RB
Add to Cart
Add to Cart

ELISA

Human NTB-A/SLAMF6 quantitates human NTB-A/SLAMF6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NTB-A/SLAMF6.

Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.05 to 10 ng/mL
Assay Sensitivity 0.05 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 114836
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Ly-108
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.