Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Obesity Panel Luminex™ Performance Control
SDP

Catalog No. QC13
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
QC13 Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC13 Supplier R&D Systems™ Supplier No. QC13

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the Human Obesity Panel

  • For use as quantitative controls for the determination of human cytokine concentrations in biological fluids
  • Concentrations have been assigned using R&D Systems' Luminex™ Performance Assays
  • Controls are prepared in a buffered protein solution with preservatives
  • They contain recombinant human cytokines at low, medium and high concentrations
  • Controls are supplied lyophilized

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Quantitative controls for the determination of human cytokine concentrations in biological fluids
Stability Stable for at least 6 months from date of receipt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.