Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OPHN1 Full-length ORF (NP_002538.1, 1 a.a. - 802 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-040 10 ug
89-955-041 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-955-040 Supplier Abnova™ Supplier No. H00004983P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a Rho-GTPase-activating protein that promotes GTP hydrolysis of Rho subfamily members. Rho proteins are important mediators of intracellular signal transduction, which affects cell migration and cell morphogenesis. Mutations in this gene are responsible for OPHN1-related X-linked mental retardation with cerebellar hypoplasia and distinctive facial dysmorhphism. [provided by RefSeq]

Sequence: MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES

Specifications

Accession Number NP_002538.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4983
Molecular Weight (g/mol) 118kDa
Name OPHN1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MRX60/OPN1
Common Name OPHN1
Gene Symbol OPHN1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.