Learn More
Abnova™ Human OR10H2 Full-length ORF (NP_039227.1, 1 a.a. - 315 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_039227.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 26538 |
| Molecular Weight (g/mol) | 61.1kDa |
| Name | OR10H2 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MLGLNHTSMSEFILVGFSAFPHLQLMLFLLFLLMYLFTLLGNLLIMATVWSERSLHTPMYLFLCVLSVSEILYTVAIIPRMLADLLSTQRSIAFLACASQMFFSFSFGFTHSFLLTVMGYDRYVAICHPLRYNVLMSPRGCACLVGCSWAGGSVMGMVVTSAIFQLTFCGSHEIQHFLCHVPPLLKLACGNNVPAVALGVGLVCIMALLGCFLLILLSYAFIVADILKIPSAEGRNKAFSTCASHLIVVIVHYGFASVIYLKPKGPHSQEGDTLMATTYAVLTPFLSPIIFSLRNKELKVAMKRTFLSTLYSSGT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.