Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OR1A2 Full-length ORF (NP_036484.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-094 25 μg
89-955-093 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-955-094 Supplier Abnova™ Supplier No. H00026189P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MKKENQSFNLDFILLGVTSQQEQNNVFFVIFLCIYPITLTGNLLIILAICADIRLHNPMYFLLANLSLVDIIFSSVTIPKVLANHLLGSKFISFGGCLMQMYFMIALAKADSYTLAAMAYDRAVAISCPLHYTTIMSPRSCILLIAGSWVIGNTSALPHTLLTASLSFCGNQEVANFYCDIMPLLKLSCSDVHFNVKMMYLGVGVFSLPLLCIIVSYVQVFSTVFQVPSTKSLFKAFCTCGSHLTVVFLYYGTTMGMYFRPLTSYSPKDAVITVMYVAVTPALNPFIYSLRNWDMKAALQKLFSKRISS

Specifications

Accession Number NP_036484.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26189
Molecular Weight (g/mol) 60.8kDa
Name OR1A2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKKENQSFNLDFILLGVTSQQEQNNVFFVIFLCIYPITLTGNLLIILAICADIRLHNPMYFLLANLSLVDIIFSSVTIPKVLANHLLGSKFISFGGCLMQMYFMIALAKADSYTLAAMAYDRAVAISCPLHYTTIMSPRSCILLIAGSWVIGNTSALPHTLLTASLSFCGNQEVANFYCDIMPLLKLSCSDVHFNVKMMYLGVGVFSLPLLCIIVSYVQVFSTVFQVPSTKSLFKAFCTCGSHLTVVFLYYGTTMGMYFRPLTSYSPKDAVITVMYVAVTPALNPFIYSLRNWDMKAALQKLFSKRISS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC119930/MGC119931/OR17-6
Common Name OR1A2
Gene Symbol OR1A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.