Learn More
Sino Biological Human Oncostatin M/OSM Protein (His & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological 10452H49HB100
Encompass_Preferred
A DNA sequence encoding the Human OSM (NP_065391.1) (Met1-Arg221) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5296
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.