Learn More
Kingfisher Biotech Inc Human Osteopontin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1551H500
Encompass_Preferred
The Human Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Human Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Human Osteopontin Specifications: (Molecular Weight: 33.7 kDa) (Amino Acid Sequence: IPVKQADSGS SEEKQLYNKY PDAVATWLNP DPSQKQNLLA PQNAVSSEET NDFKQETLPS KSNESHDHMD DMDDEDDDDH VDSQDSIDSN DSDDVDDTDD SHQSDESHHS DESDELVTDF PTDLPATEVF TPVVPTVDTY DGRGDSVVYG LRSKSKKFRR PDIQYPDATD EDITSHMESE ELNGAYKAIP VAQDLNAPSD WDSRGKDSYE TSQLDDQSAE THSHKQSRLY KRKANDESNE HSDVIDSQEL SKVSREFHSH EFHSHEDMLV VDPKSKEEDK HLKFRISHEL DSASSEVN (298)) (Gene ID: 6696). For research use only. Made in the USA
Catalog No. 50-253-3151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.