Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Osteopontin/OPN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067857
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB14331SP 25 μg
MAB14331100 100 μg
MAB14331500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB14331SP Supplier R&D Systems Supplier No. MAB14331SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Osteopontin/OPN
Applications Western Blot
Classification Monoclonal
Clone 223126
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 100 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000573.1
Gene Alias BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Gene Symbols SPP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Osteopontin Ile17-Asn300 Accession # NP_000573.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6696
Test Specificity Detects human Osteopontin in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Osteopontin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.