Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Osteoprotegerin (TNFRSF11B) Recombinant Protein, PeproTech®

Catalog No. 45014100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45014100UG 100 μg
450-14-10UG 10 μg
450-14-1MG 1 mg
45014250UG 250 μg
45014500UG 500 μg
450-14-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45014100UG Supplier Gibco™ Supplier No. 45014100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human OPG is a soluble 20.0 kDa protein containing 174 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00300
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4982
Molecular Weight (g/mol) 20 kDa
Name Human Osteoprotegerin (TNFRSF11B)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11 B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11 b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11 B; tumor necrosis factor receptor superfamily, member 11 b; tumor necrosis factor receptor superfamily, member 11 b (osteoprotegerin)
Common Name TNFRSF11B
Gene Symbol TNFRSF11B
Biological Activity Determined by its ability to inhibit TRAIL-induced apoptosis of LN-18 glioblastoma cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence METFPPKYLH YDEETSHQLL CDKCPPGTYL KQHCTAKWKT VCAPCPDHYY TDSWHTSDEC LYCSPVCKEL QYVKQECNRT HNRVCECKEG RYLEIEFCLK HRSCPPGFGV VQAGTPERNT VCKRCPDGFF SNETSSKAPC RKHTNCSVFG LLLTQKGNAT HDNICSGNSE STQK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.